Assay Details
Target Gene Details
Entrez Gene ID: | 67863 |
Gene Name: | solute carrier family 25 (mitochondrial carrier oxoglutarate carrier), member 11 |
Gene Aliases: |
2310022P18Rik, 2oxoc |
Location: |
Chr.11:70644019-70647423 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a11 | NM_024211.3 | 8 | 1450 | NP_077173.1 |
| AI196845.1 | 1 | 420 | ||
| AK003957.1 | 7 | 1089 | BAB23092.1 | |
| AK009487.1 | 8 | 1396 | BAB26319.1 | |
| AK009824.1 | 8 | 1350 | BAB26524.1 | |
| AK168951.1 | 8 | 1375 | BAE40757.1 | |
| AK178451.1 | 1 | 188 | ||
| AK206341.1 | 1 | 89 | ||
| AK210712.1 | 1 | 188 | ||
| AK212009.1 | 1 | 157 | ||
| AK214004.1 | 1 | 188 | ||
| BC003455.1 | 8 | 1426 | AAH03455.1 | |
| BC019631.1 | 8 | 1404 | AAH19631.1 |
More Information
Set Membership: |
|

Genomic Map