Assay Details
Target Gene Details
Entrez Gene ID: | 67283 |
Gene Name: | solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19 |
Gene Aliases: |
2900089E13Rik, DNC, MUP1, TPC |
Location: |
Chr.11:115614170-115628295 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a19 | NM_001252384.1 | 8 | 1308 | NP_001239313.1 |
| NM_001252394.1 | 6 | 961 | NP_001239323.1 | |
| NM_001252395.1 | 8 | 1320 | NP_001239324.1 | |
| NM_001252396.1 | 6 | 975 | NP_001239325.1 | |
| NM_026071.3 | 7 | 1145 | NP_080347.1 | |
| AK005710.1 | 7 | 990 | BAB24199.1 | |
| AK168145.1 | 6 | 760 | ||
| AK172622.1 | 6 | 947 | BAE43100.1 | |
| BC018167.1 | 7 | 951 | AAH18167.1 | |
| BC046767.1 | 7 | 995 | AAH46767.1 | |
| CF730205.1 |
More Information
Set Membership: |
|

Genomic Map