Assay Details
Target Gene Details
Entrez Gene ID: | 68497 |
Gene Name: | apoptosis resistant E3 ubiquitin protein ligase 1 |
Gene Aliases: |
1110018G07Rik, AI649076, AW610792, Kiaa0317, mKIAA0317 |
Location: |
Chr.12:84918140-84970886 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arel1 | NM_178065.4 | 2 | 377 | NP_835166.3 |
| AB093230.1 | BAC41414.1 | |||
| AK147491.1 | 2 | 378 | ||
| AK147878.1 | 2 | 341 | BAE28198.1 | |
| AK154529.1 | 2 | 358 | BAE32654.1 | |
| CJ128427.1 | 2 | 378 |
More Information
Set Membership: |
|

Genomic Map