Assay Details
Target Gene Details
Entrez Gene ID: | 11687 |
Gene Name: | arachidonate 15-lipoxygenase |
Gene Aliases: |
12-LO, 12/15-LO, 15-LOX, Alox12l, L-12LO |
Location: |
Chr.11:70344147-70375918 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alox15 | NM_009660.3 | 14 | 2122 | NP_033790.3 |
| AK142371.1 | 14 | 2123 | BAE25045.1 | |
| BC056625.1 | 14 | 2109 | AAH56625.1 | |
| BC081546.1 | 14 | 2108 | AAH81546.1 | |
| L34570.1 | 14 | 2106 | AAA64930.1 |
More Information
Set Membership: |
|

Genomic Map