Assay Details
Target Gene Details
Entrez Gene ID: | 16834 |
Gene Name: | component of oligomeric golgi complex 1 |
Gene Aliases: |
6030445I15, Ldlb, mKIAA1381 |
Location: |
Chr.11:113649178-113669331 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cog1 | NM_013581.3 | 1 | 106 | NP_038609.3 |
| AF109377.2 | 1 | 89 | AAD13780.2 | |
| AK034636.2 | 1 | 111 | BAE43288.1 | |
| AK152464.1 | 1 | 97 | BAE31241.1 | |
| AK158507.1 | 1 | 467 | BAE34539.1 | |
| AK159497.1 | 1 | 108 | BAE35130.1 | |
| AK162971.1 | 1 | 107 | BAE37141.1 | |
| AK171574.1 | 1 | 100 | BAE42534.1 | |
| AK220387.1 | 1 | 103 | BAD90252.1 | |
| BC049541.1 | 1 | 307 | AAH49541.1 | |
| BC063056.1 | 1 | 88 | AAH63056.1 | |
| BY721599.1 | 1 | 107 |
Target Gene Details
Entrez Gene ID: | 69806 |
Gene Name: | solute carrier family 39 (metal ion transporter), member 11 |
Gene Aliases: |
1810074D23Rik |
Location: |
Chr.11:113244851-113650752 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a11 | AK165545.1 |

Genomic Map