Assay Details
Target Gene Details
Entrez Gene ID: | 13866 |
Gene Name: | v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) |
Gene Aliases: |
Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023 |
Location: |
Chr.11:98412484-98437716 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erbb2 | NM_001003817.1 | 26 | 3507 | NP_001003817.1 |
| AK031099.1 | 26 | 3375 | ||
| AK031542.1 | 18 | 2205 | BAC27442.1 | |
| AK083669.1 | 25 | 3457 | ||
| AK129487.1 | 26 | 3477 | BAC98297.1 | |
| AK142296.1 | 26 | 3501 | ||
| BC027080.1 | 8 | 1119 | ||
| BC046811.1 | 25 | 3579 | AAH46811.1 | |
| BC053078.1 | 25 | 3579 | AAH53078.1 | |
| L47239.1 | AAA93532.1 |
More Information
Set Membership: |
|

Genomic Map