Assay Details
Target Gene Details
Entrez Gene ID: | 238023 |
Gene Name: | hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing |
Gene Aliases: |
BC069960 |
Location: |
Chr.11:121204433-121222656 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hexdc | NM_001001333.2 | 13 | 1796 | NP_001001333.2 |
| NM_001146073.1 | 12 | 1601 | NP_001139545.1 | |
| AI481765.1 | ||||
| AK154237.1 | 13 | 1770 | BAE32455.1 | |
| BC069960.1 | 12 | 1553 | AAH69960.1 | |
| FM204885.1 | 12 | 1333 | ||
| FM204886.1 | 12 | 1552 |

Genomic Map