Assay Details
Target Gene Details
Entrez Gene ID: | 68876 |
Gene Name: | XRCC6 binding protein 1 |
Gene Aliases: |
1110068E08Rik, 2410012H02Rik |
Location: |
Chr.10:126868427-126901711 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc6bp1 | NM_001159559.1 | 1 | 226 | NP_001153031.1 |
| NM_026858.3 | 1 | 226 | NP_081134.2 | |
| AK004405.1 | 1 | 206 | ||
| AK010471.1 | 1 | 210 | BAB26964.1 | |
| AK039002.1 | 1 | 206 | ||
| BC030630.1 | 1 | 188 | AAH30630.1 | |
| BG801921.1 | 1 | 226 |
More Information
Set Membership: |
|

Genomic Map