Assay Details
Target Gene Details
Entrez Gene ID: | 320655 |
Gene Name: | post-GPI attachment to proteins 3 |
Gene Aliases: |
CAB2, D430035D22Rik, Perld1 |
Location: |
Chr.11:98388672-98400539 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pgap3 | NM_001033537.2 | 8 | 1944 | NP_001028709.2 |
| AK052486.1 | 8 | 1861 | ||
| AK078974.1 | 8 | 1870 | ||
| AK170817.1 | 8 | 1859 | BAE42049.1 | |
| BC030368.1 | 6 | 1564 | AAH30368.1 |

Genomic Map