Assay Details
Target Gene Details
Entrez Gene ID: | 215113 |
Gene Name: | solute carrier family 43, member 2 |
Gene Aliases: |
7630402D21Rik, BC042513, Lat4 |
Location: |
Chr.11:75530282-75577575 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc43a2 | NM_001199283.1 | 14 | 3698 | NP_001186212.1 |
| NM_001199284.1 | 14 | 3480 | NP_001186213.1 | |
| NM_173388.2 | 14 | 3443 | NP_775564.1 | |
| AK143773.1 | 14 | 3541 | BAE25533.1 | |
| AK158440.1 | 5 | 2419 | BAE34511.1 | |
| BC042513.1 | 14 | 3439 | AAH42513.1 |

Genomic Map