Assay Details
Target Gene Details
Entrez Gene ID: | 66272 |
Gene Name: | cytochrome c oxidase assembly protein 16 |
Gene Aliases: |
1810020G14Rik, 1810055I05Rik, BB388670 |
Location: |
Chr.12:81470704-81485110 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox16 | NM_025461.5 | 4 | 1180 | NP_079737.1 |
| AK082374.1 | 4 | 1181 | BAC38480.1 |

Genomic Map