Assay Details
Target Gene Details
Entrez Gene ID: | 237940 |
Gene Name: | amine oxidase, copper containing 2 (retina-specific) |
Gene Aliases: |
RAO |
Location: |
Chr.11:101325063-101329699 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aoc2 | NM_178932.1 | 1 | 1352 | NP_849263.1 |
| AF350445.2 | 1 | 1352 | AAK58864.2 | |
| BC150843.1 | 1 | 1343 | ||
| BC150848.1 | 1 | 1343 |
More Information
Set Membership: |
|

Genomic Map