Assay Details
Target Gene Details
Entrez Gene ID: | 53413 |
Gene Name: | exocyst complex component 7 |
Gene Aliases: |
Exo70, sec70 |
Location: |
Chr.11:116287998-116306749 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exoc7 | NM_001162872.1 | 18 | 2785 | NP_001156344.1 |
| NM_016857.2 | 20 | 2917 | NP_058553.2 | |
| AK140450.1 | 20 | 2891 | BAE24393.1 |
More Information
Set Membership: |
|

Genomic Map