Assay Details
Target Gene Details
Entrez Gene ID: | 432552 |
Gene Name: | family with sequence similarity 71, member B |
Gene Aliases: |
Gari-l3, OTTMUSG00000005491 |
Location: |
Chr.11:46404728-46407985 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam71b | NM_001013783.2 | 1 | 581 | NP_001013805.1 |
| AB473727.1 | 1 | 506 | ||
| BB615215.1 | 1 | 582 | ||
| BC150988.1 | 1 | 534 |

Genomic Map