Assay Details
Target Gene Details
Entrez Gene ID: | 16882 |
Gene Name: | ligase III, DNA, ATP-dependent |
Gene Aliases: |
D11Wsu78e |
Location: |
Chr.11:82781109-82804274 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lig3 | NM_001291245.1 | 2 | 253 | NP_001278174.1 |
| NM_001291246.1 | 2 | 253 | NP_001278175.1 | |
| NM_001291247.1 | 2 | 257 | NP_001278176.1 | |
| NM_010716.3 | 2 | 257 | NP_034846.2 | |
| AK010719.1 | 2 | 168 | ||
| AK031540.1 | 2 | 195 | ||
| AK143531.1 | 2 | 123 | BAE25421.1 | |
| AK150644.1 | 2 | 165 | BAE29732.1 | |
| AK156018.1 | 2 | 175 | BAE33548.1 | |
| AK167363.1 | 2 | 165 | BAE39460.1 | |
| BC049240.1 | 2 | 160 | AAH49240.1 | |
| BC083500.1 | 2 | 156 | AAH83500.1 | |
| BY241147.1 | 2 | 231 | ||
| BY307824.1 | 2 | 257 | ||
| U66057.1 | 2 | 182 | AAC53003.1 | |
| U66058.1 | 2 | 182 | AAC53004.1 |
More Information
Set Membership: |
|

Genomic Map