Assay Details
Target Gene Details
Entrez Gene ID: | 16882 |
Gene Name: | ligase III, DNA, ATP-dependent |
Gene Aliases: |
D11Wsu78e |
Location: |
Chr.11:82781109-82804274 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lig3 | NM_001291245.1 | 20 | 5460 | NP_001278174.1 |
| NM_001291246.1 | 20 | 5457 | NP_001278175.1 | |
| NM_001291247.1 | 20 | 5461 | NP_001278176.1 | |
| NM_010716.3 | 20 | 5464 | NP_034846.2 | |
| AK156030.1 | 9 | 3446 | BAE33553.1 | |
| AK156825.1 | 9 | 3446 | BAE33868.1 | |
| AK169648.1 | 1 | 2198 | ||
| BC083500.1 | 20 | 5363 | AAH83500.1 | |
| CF198126.1 | ||||
| W83057.1 | 1 | 468 |
More Information
Set Membership: |
|

Genomic Map