Assay Details
Target Gene Details
Entrez Gene ID: | 118453 |
Gene Name: | matrix metallopeptidase 28 (epilysin) |
Gene Aliases: |
D130023P12 |
Location: |
Chr.11:83441241-83463064 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp28 | NM_080453.2 | 8 | 2133 | NP_536701.1 |
| NM_172797.2 | 8 | 2091 | NP_766385.1 | |
| AK051258.1 | 8 | 2154 | BAC34577.1 | |
| AY065653.1 | 8 | 2163 | AAL47576.1 | |
| AY071828.1 | 8 | 2133 | AAL57761.1 | |
| AY071829.1 | 8 | 2091 | AAL57762.1 |
More Information
Set Membership: |
|

Genomic Map