Assay Details
Target Gene Details
Entrez Gene ID: | 17428 |
Gene Name: | max binding protein |
Gene Aliases: |
Rox, bHLHd3 |
Location: |
Chr.11:74830796-74845725 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mnt | NM_010813.3 | 2 | 582 | NP_034943.3 |
| AK147933.1 | 2 | 566 | BAE28239.1 | |
| BC054534.1 | 2 | 508 | AAH54534.1 | |
| CA327080.1 | 2 | 586 | ||
| U77356.1 | 2 | 582 | AAB38687.1 | |
| Y07609.1 | 2 | 528 | CAA68878.1 |
More Information
Set Membership: |
|

Genomic Map