Assay Details
Target Gene Details
Entrez Gene ID: | 17428 |
Gene Name: | max binding protein |
Gene Aliases: |
Rox, bHLHd3 |
Location: |
Chr.11:74830796-74845725 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mnt | NM_010813.3 | 6 | 4047 | NP_034943.3 |
| AK014288.1 | 1 | 309 | ||
| AK147933.1 | 6 | 4031 | BAE28239.1 | |
| AK219563.1 | 1 | 40 | ||
| BC054534.1 | 6 | 3973 | AAH54534.1 | |
| U77356.1 | 6 | 4047 | AAB38687.1 |
More Information
Set Membership: |
|

Genomic Map