Assay Details
Target Gene Details
Entrez Gene ID: | 19193 |
Gene Name: | pipecolic acid oxidase |
Gene Aliases: |
LPIPOX, Pso |
Location: |
Chr.11:77880614-77893872 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pipox | NM_008952.2 | 8 | 1566 | NP_032978.2 |
| AK008555.2 | 8 | 1527 | BAB25741.1 | |
| AK149488.1 | 8 | 1515 | BAE28913.1 | |
| BC013525.1 | 8 | 1480 | AAH13525.1 | |
| U94700.1 | 8 | 1478 | AAC39948.1 |
More Information
Set Membership: |
|

Genomic Map