Assay Details
Target Gene Details
Entrez Gene ID: | 18738 |
Gene Name: | phosphatidylinositol transfer protein, alpha |
Gene Aliases: |
Pitpn, vb, vibrator |
Location: |
Chr.11:75588108-75628778 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitpna | NM_008850.1 | 11 | 2397 | NP_032876.1 |
| AK144618.1 | 12 | 2410 | BAE25972.1 | |
| AK165659.1 | 12 | 2402 | BAE38321.1 | |
| AK168720.1 | 12 | 2400 | BAE40562.1 | |
| U96725.1 | 11 | 2398 | AAC53266.1 |
More Information
Set Membership: |
|

Genomic Map