Assay Details
Target Gene Details
Entrez Gene ID: | 71795 |
Gene Name: | phosphatidylinositol transfer protein, cytoplasmic 1 |
Gene Aliases: |
1110020B03Rik, 5830436L09Rik, AI662802, AI851387, C330017I21Rik, Dnr411, RDGBB, RDGBB1, rdgB-beta |
Location: |
Chr.11:107207892-107470892 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitpnc1 | NM_145823.2 | 10 | 4773 | NP_665822.1 |
| AK034285.1 | 1 | 2328 | ||
| AK043007.1 | 1 | 272 | ||
| AK043272.1 | 1 | 272 | ||
| AK153793.1 | 1 | 1121 | BAE32184.1 | |
| AK163426.1 | 1 | 2330 | BAE37344.1 |
More Information
Set Membership: |
|

Genomic Map