Assay Details
Target Gene Details
Entrez Gene ID: | 12351 |
Gene Name: | carbonic anhydrase 4 |
Gene Aliases: |
AW456718, Ca4 |
Location: |
Chr.11:84957754-84966054 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car4 | NM_007607.2 | 8 | 1075 | NP_031633.1 |
| AK154754.1 | 8 | 1041 | BAE32805.1 | |
| AK165399.1 | 8 | 1004 | ||
| BC012704.1 | 8 | 1000 | AAH12704.1 | |
| BM935015.1 | 2 | 329 |

Genomic Map