Assay Details
Target Gene Details
Entrez Gene ID: | 14104 |
Gene Name: | fatty acid synthase |
Gene Aliases: |
A630082H08Rik, FAS |
Location: |
Chr.11:120805958-120824547 on Build GRCm38 |
Assay Gene Location: | Within Exon 43 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fasn | NM_007988.3 | 43 | 9263 | NP_032014.3 |
AK078721.1 | 1 | 691 | ||
AK080374.1 | 17 | 4160 | BAC37895.1 |
