Assay Details
Target Gene Details
Entrez Gene ID: | 83796 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 |
Gene Aliases: |
AW322457, Baf60b |
Location: |
Chr.11:106263171-106272972 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarcd2 | NM_001130187.1 | 13 | 2593 | NP_001123659.1 |
| NM_031878.2 | 12 | 2227 | NP_114084.2 | |
| AK159259.1 | 13 | 2430 | BAE34940.1 | |
| AV317359.1 | 1 | 68 | ||
| AV367494.1 | 1 | 124 | ||
| BC005732.1 | 12 | 2217 | AAH05732.1 | |
| BE853586.1 | 1 | 175 |
More Information
Set Membership: |
|

Genomic Map