Assay Details
Target Gene Details
Entrez Gene ID: | 68067 |
Gene Name: | RIKEN cDNA 3010026O09 gene |
Gene Aliases: |
2900016G09Rik, AI851514, AW475975 |
Location: |
Chr.11:50174851-50200115 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3010026O09Rik | NM_026543.3 | 7 | 1038 | NP_080819.2 |
| AK003636.1 | 7 | 1013 | BAB22904.1 | |
| AK019392.1 | 7 | 1039 | BAB31698.1 | |
| BC028428.1 | 7 | 1023 | AAH28428.1 |

Genomic Map