Assay Details
Target Gene Details
Entrez Gene ID: | 432572 |
Gene Name: | sperm antigen with calponin homology and coiled-coil domains 1 |
Gene Aliases: |
2810012G08Rik, B230396K10Rik, Cytsb |
Location: |
Chr.11:61956763-62223013 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Specc1 | NM_001029936.3 | 13 | 5509 | NP_001025107.1 |
| NM_001281818.1 | 15 | 5726 | NP_001268747.1 | |
| AK012725.1 | 1 | 571 | ||
| AK220465.1 | 13 | 5436 | BAD90492.1 |

Genomic Map