Assay Details
Target Gene Details
Entrez Gene ID: | 50529 |
Gene Name: | mitchondrial ribosomal protein S7 |
Gene Aliases: |
MRP-S7, Rpms7 |
Location: |
Chr.11:115604151-115607624 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps7 | NM_025305.2 | 5 | 1056 | NP_079581.1 |
| AK009251.1 | 5 | 1021 | BAB26169.1 | |
| AK012225.1 | ||||
| AK131937.1 | BAE20885.1 | |||
| AK162024.1 | 5 | 1035 | BAE36686.1 | |
| BC049636.1 | 5 | 1056 | AAH49636.1 |
More Information
Set Membership: |
|

Genomic Map