Assay Details
Target Gene Details
Entrez Gene ID: | 11484 |
Gene Name: | aspartoacylase |
Gene Aliases: |
Acy-2, Acy2, nur7 |
Location: |
Chr.11:73304988-73326868 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aspa | NM_023113.5 | 1 | 318 | NP_075602.2 |
| AF212998.1 | 1 | 168 | AAF76459.1 | |
| AI527854.1 | 1 | 318 | ||
| AK002859.1 | 1 | 316 | BAB22412.1 | |
| AK008354.1 | 1 | 321 | BAB25624.1 | |
| AK035910.1 | BAC29240.1 | |||
| BC024934.1 | 1 | 282 | AAH24934.1 |

Genomic Map