Assay Details
Target Gene Details
Entrez Gene ID: | 13169 |
Gene Name: | drebrin-like |
Gene Aliases: |
Abp1, SH3P7, mAbp1 |
Location: |
Chr.11:5788483-5800980 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbnl | NM_001146308.1 | 13 | 1426 | NP_001139780.1 |
| NM_001146309.1 | 13 | 1414 | NP_001139781.1 | |
| NM_013810.3 | 13 | 1417 | NP_038838.1 | |
| AK046073.1 | 13 | 1394 | BAC32592.1 | |
| AK053796.1 | 13 | 1410 | BAC35528.1 | |
| AK078082.1 | 13 | 1381 | BAC37118.1 | |
| AK134136.1 | 13 | 1381 | BAE22028.1 | |
| AK134487.1 | 13 | 1393 | BAE22160.1 | |
| AK140752.1 | 13 | 1384 | BAE24466.1 | |
| AK142818.1 | 13 | 1364 | BAE25201.1 | |
| AK146920.1 | 13 | 1381 | BAE27532.1 | |
| AK151096.1 | 13 | 1394 | BAE30108.1 | |
| AK152110.1 | 13 | 1382 | ||
| AK152204.1 | 13 | 1394 | BAE31033.1 | |
| AK153072.1 | 13 | 1393 | ||
| AK153389.1 | 13 | 1371 | BAE31953.1 | |
| AK159984.1 | 13 | 1391 | BAE35535.1 | |
| AK161973.1 | 12 | 1688 | ||
| AK167956.1 | 13 | 1353 | ||
| AK168898.1 | 13 | 1391 | BAE40714.1 | |
| AK172471.1 | 13 | 1371 | BAE43024.1 | |
| AK190211.1 | 1 | 57 | ||
| BC046430.1 | 13 | 1379 | AAH46430.1 | |
| U58884.1 | 13 | 1370 | AAC52640.1 |
More Information
Set Membership: |
|

Genomic Map