Assay Details
Target Gene Details
Entrez Gene ID: | 98238 |
Gene Name: | leucine rich repeat containing 59 |
Gene Aliases: |
AA959742, C78668 |
Location: |
Chr.11:94629824-94645216 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc59 | NM_133807.1 | 7 | 1177 | NP_598568.1 |
| AK053450.1 | 7 | 1204 | BAC35390.1 | |
| AK150916.1 | 2 | 489 | BAE29954.1 | |
| AK159374.1 | 7 | 1233 | BAE35030.1 | |
| AK159552.1 | 7 | 1227 | BAE35177.1 | |
| AK166576.1 | 7 | 1226 | BAE38865.1 | |
| AK167604.1 | 7 | 1168 | BAE39660.1 | |
| AK169250.1 | 5 | 789 | BAE41014.1 | |
| AK169390.1 | 7 | 1176 | BAE41137.1 | |
| AK206116.1 | ||||
| AK212896.1 | 1 | 56 | ||
| AK213336.1 | ||||
| BC006877.1 | 7 | 1177 | AAH06877.1 | |
| BC066172.1 | 7 | 1171 | AAH66172.1 |
More Information
Set Membership: |
|

Genomic Map