Assay Details
Target Gene Details
Entrez Gene ID: | 50724 |
Gene Name: | SAP30-like |
Gene Aliases: |
2310079P12Rik, AF006998, L55 |
Location: |
Chr.11:57801637-57810615 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap30l | NM_001081168.1 | 1 | 538 | NP_001074637.1 |
| AK010234.1 | BAB26785.1 | |||
| BC051686.1 | 1 | 538 | ||
| BC094930.1 | 2 | 257 | AAH94930.1 | |
| BC117029.1 | 1 | 252 | ||
| BC117031.1 | 1 | 252 |
More Information
Set Membership: |
|

Genomic Map