Assay Details
Target Gene Details
Entrez Gene ID: | 238330 |
Gene Name: | interferon regulatory factor 2 binding protein-like |
Gene Aliases: |
6430527G18Rik, Eap1 |
Location: |
Chr.12:86880703-86884814 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf2bpl | NM_145836.2 | 1 | 2059 | NP_665835.1 |
| AF525300.1 | 1 | 2056 | AAM82165.1 | |
| AK032366.1 | 1 | 2064 | BAC27837.1 | |
| AK032622.1 | 1 | 2063 | BAC27955.1 | |
| AK082791.1 | 1 | 2043 | BAC38621.1 | |
| AK084782.1 | 1 | 2059 | ||
| AK140441.1 | 1 | 2040 | BAE24388.1 | |
| AK154383.1 | 1 | 1808 | BAE32550.1 | |
| AK155373.1 | 1 | 2013 | BAE33226.1 | |
| AK160059.1 | 1 | 2013 | ||
| AK171043.1 | 1 | 2013 | BAE42208.1 | |
| AK173282.1 | 1 | 2059 | BAD32560.1 | |
| AK203324.1 | ||||
| BC057128.1 | 1 | 1496 | AAH57128.1 | |
| BC063253.1 | 1 | 1496 | AAH63253.1 |
More Information
Set Membership: |
|

Genomic Map