Assay Details
Target Gene Details
Entrez Gene ID: | 16191 |
Gene Name: | interleukin 5 |
Gene Aliases: |
Il-5 |
Location: |
Chr.11:53719364-53725623 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il5 | NM_010558.1 | 4 | 542 | NP_034688.1 |
| BC125366.1 | 4 | 527 | ||
| BC132066.1 | 4 | 527 | ||
| X04601.1 | 4 | 542 | CAA28266.1 | |
| X06270.1 | 4 | 542 | CAA29606.1 |

Genomic Map