Assay Details
Target Gene Details
Entrez Gene ID: | 66048 |
Gene Name: | ER membrane protein complex subunit 6 |
Gene Aliases: |
0610009E20Rik, 0610025L18Rik, Tmem93 |
Location: |
Chr.11:73175503-73177042 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc6 | NM_001168470.1 | 2 | 1172 | NP_001161942.1 |
| NM_025318.3 | 2 | 1123 | NP_079594.1 | |
| AK002385.1 | 2 | 1109 | BAB22059.1 | |
| AK003198.1 | 2 | 1119 | BAB22636.1 | |
| AK012769.1 | 2 | 1109 | BAB28457.1 | |
| AK166490.1 | 1 | 395 | ||
| BC022104.1 | 2 | 1094 | AAH22104.1 |

Genomic Map