Assay Details
Target Gene Details
Entrez Gene ID: | 17762 |
Gene Name: | microtubule-associated protein tau |
Gene Aliases: |
AI413597, AW045860, Mtapt, Tau |
Location: |
Chr.11:104231353-104332090 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapt | NM_001038609.2 | 12 | 1946 | NP_001033698.1 |
| NM_001285454.1 | NP_001272383.1 | |||
| NM_001285455.1 | 9 | 1706 | NP_001272384.1 | |
| NM_001285456.1 | 9 | 1679 | NP_001272385.1 | |
| NM_010838.4 | 10 | 1772 | NP_034968.3 | |
| AK147636.1 | 12 | 1907 | BAE28038.1 | |
| BM950831.1 | ||||
| M18776.1 | AAA40166.1 |

Genomic Map