Assay Details
Target Gene Details
Entrez Gene ID: | 380683 |
Gene Name: | SEC14-like 3 (S. cerevisiae) |
Gene Aliases: |
1110069O07Rik |
Location: |
Chr.11:4064853-4078990 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec14l3 | NM_001029937.2 | 12 | 1252 | NP_001025108.1 |
| AK136060.1 | 6 | 605 | BAE22802.1 | |
| BC138982.1 | 12 | 1252 | ||
| BC138985.1 | 12 | 1252 |

Genomic Map