Assay Details
Target Gene Details
Entrez Gene ID: | 327959 |
Gene Name: | XIAP associated factor 1 |
Gene Aliases: |
Fbox39 |
Location: |
Chr.11:72301436-72313733 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xaf1 | NM_001037713.4 | 6 | 1405 | NP_001032802.2 |
| NM_001291153.1 | 4 | 1120 | NP_001278082.1 |

Genomic Map