Assay Details
Target Gene Details
Entrez Gene ID: | 56622 |
Gene Name: | a disintegrin and metallopeptidase domain 21 |
Gene Aliases: |
Adam31 |
Location: |
Chr.12:81558584-81568474 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam21 | NM_020330.4 | 1 | 255 | NP_065063.1 |
| AK014827.2 | 1 | 256 | BAB29569.1 |
More Information
Set Membership: |
|

Genomic Map