Assay Details
Target Gene Details
Entrez Gene ID: | 107817 |
Gene Name: | jumonji domain containing 6 |
Gene Aliases: |
5730436I23Rik, D11Ertd195e, PSR, PtdSerR, Ptdsr, mKIAA0585 |
Location: |
Chr.11:116837432-116843449 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jmjd6 | NM_033398.2 | 6 | 1396 | NP_203971.2 |
| AF304118.1 | 6 | 1329 | AAG27719.1 | |
| AK122317.1 | 3 | 4267 | BAC65599.1 | |
| BC056629.1 | 6 | 1396 | AAH56629.1 | |
| BC117737.1 | 6 | 1131 | ||
| BP769727.1 | 1 | 293 | ||
| EF527404.1 | 7 | 1485 | ||
| EF527405.1 | 7 | 1233 |
More Information
Set Membership: |
|

Genomic Map