Assay Details
Target Gene Details
Entrez Gene ID: | 360013 |
Gene Name: | myosin XVIIIA |
Gene Aliases: |
MAJN, MyoPDZ, MysPDZ |
Location: |
Chr.11:77763251-77865988 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myo18a | NM_001291212.1 | 1 | 727 | NP_001278141.1 |
| NM_001291213.1 | 1 | 727 | NP_001278142.1 | |
| NM_011586.3 | 1 | 727 | NP_035716.1 | |
| AB026497.1 | 1 | 697 | BAA93660.1 | |
| AK146852.1 | 2 | 807 | BAE27482.1 | |
| BC138366.1 | 1 | 669 |

Genomic Map