Assay Details
Target Gene Details
Entrez Gene ID: | 70310 |
Gene Name: | phospholipid scramblase 3 |
Gene Aliases: |
2210403O21Rik, 2610037N06Rik, ESTM3, X83310 |
Location: |
Chr.11:69846251-69852058 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plscr3 | NM_001168497.1 | 8 | 2310 | NP_001161969.1 |
| NM_023564.4 | 8 | 2234 | NP_076053.1 | |
| NR_033133.1 | 7 | 2423 | ||
| AK029154.1 | 7 | 2390 | BAC26329.1 | |
| AK172635.1 | 8 | 2211 | BAE43107.1 |
Target Gene Details
Entrez Gene ID: | 83813 |
Gene Name: | tyrosine kinase, non-receptor, 1 |
Gene Aliases: |
Kos1, Tnk1a, Tnk1b |
Location: |
Chr.11:69850563-69858730 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnk1 | NM_031880.3 | 14 | 2276 | NP_114086.3 |
| AK033440.1 | 9 | 1647 | BAC28288.1 | |
| AK076220.1 | 14 | 2283 | ||
| BC029623.1 | 13 | 2263 | AAH29623.1 | |
| BC055303.1 | 13 | 2180 | AAH55303.1 |
More Information
Set Membership: |
|

Genomic Map