Assay Details
Target Gene Details
Entrez Gene ID: | 11302 |
Gene Name: | apoptosis-associated tyrosine kinase |
Gene Aliases: |
AATYK, aatyk1, mKIAA0641 |
Location: |
Chr.11:120007311-120047182 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aatk | NM_001198785.1 | 14 | 4661 | NP_001185714.1 |
| NM_001198787.1 | 14 | 4433 | NP_001185716.1 | |
| NM_007377.4 | 14 | 4944 | NP_031403.2 | |
| AB093253.2 | 14 | 4944 | BAC41437.2 | |
| AF011908.1 | 14 | 4400 | AAB71837.1 | |
| AK036705.1 | 14 | 4435 | BAC29541.1 | |
| AK076214.1 | 1 | 134 | ||
| AK147239.1 | 10 | 5284 | BAE27789.1 | |
| AK155100.1 | 14 | 4435 | BAE33045.1 | |
| AY236858.1 | 14 | 4439 | AAO92350.1 | |
| AY236859.1 | 14 | 4466 | AAO92351.1 | |
| BC080846.1 | 14 | 4515 | AAH80846.1 |
More Information
Set Membership: |
|

Genomic Map