Assay Details
Target Gene Details
Entrez Gene ID: | 20747 |
Gene Name: | speckle-type POZ protein |
Gene Aliases: |
AI315626, Pcif1, TEF2 |
Location: |
Chr.11:95414064-95493410 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spop | NM_025287.2 | 11 | 2588 | NP_079563.2 |
| AB071989.1 | 9 | 2397 | BAB68542.1 | |
| AK132383.1 | 10 | 2764 | ||
| AK159482.1 | 10 | 2741 | BAE35119.1 | |
| AK168185.1 | 10 | 2710 | BAE40145.1 | |
| CJ207806.1 | 1 | 105 |
More Information
Set Membership: |
|

Genomic Map