Assay Details
Target Gene Details
Entrez Gene ID: | 27274 |
Gene Name: | zinc finger protein 354B |
Gene Aliases: |
Kid2, Tcf17l1, Znf354b |
Location: |
Chr.11:50921786-50931635 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp354b | NM_013744.3 | 5 | 2147 | NP_038772.1 |
| AF184112.1 | 4 | 2016 | AAF01034.1 | |
| AK010007.1 | 1 | 1234 | BAB26638.1 | |
| AK148756.1 | 5 | 2149 | ||
| BC107400.1 | AAI07401.1 | |||
| BC107401.1 | AAI07402.1 |
More Information
Set Membership: |
|

Genomic Map