Assay Details
Target Gene Details
Entrez Gene ID: | 70355 |
Gene Name: | G protein-coupled receptor, family C, group 5, member C |
Gene Aliases: |
1110028I06Rik, 3200002M13Rik, Raig3 |
Location: |
Chr.11:114851146-114872617 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprc5c | NM_001110337.1 | 5 | 2065 | NP_001103807.1 |
| AK128950.1 | 5 | 1755 | ||
| CO425185.1 | 3 | 633 |
More Information
Set Membership: |
|

Genomic Map