Assay Details
Target Gene Details
Entrez Gene ID: | 14783 |
Gene Name: | growth factor receptor bound protein 10 |
Gene Aliases: |
5730571D09Rik, AI325020, Meg1, mKIAA0207 |
Location: |
Chr.11:11930499-12037422 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grb10 | NM_001177629.1 | 16 | 3197 | NP_001171100.1 |
| NM_010345.4 | 17 | 3522 | NP_034475.2 | |
| AK017862.1 | 1 | 94 | ||
| AK030727.1 | 17 | 3457 | BAC27100.1 | |
| AK160651.1 | 1 | 1048 | ||
| AK169000.1 | 10 | 2542 | BAE40799.1 | |
| BC016111.1 | 16 | 3197 | AAH16111.1 | |
| BC053842.1 | 17 | 3489 | AAH53842.1 |
More Information
Set Membership: |
|

Genomic Map