Assay Details
Target Gene Details
Entrez Gene ID: | 216858 |
Gene Name: | potassium channel tetramerisation domain containing 11 |
Gene Aliases: |
AF465352, Ren |
Location: |
Chr.11:69878264-69880985 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kctd11 | NM_153143.4 | 1 | 1822 | NP_694783.1 |
| AF465352.1 | 1 | 1789 | AAM95915.1 | |
| AK161602.1 | 1 | 1817 | BAE36486.1 | |
| AK170258.1 | 1 | 1716 | BAE41666.1 | |
| BC052502.1 | 1 | 1070 | AAH52502.1 | |
| BC052734.1 | 1 | 1773 | AAH52734.1 |
More Information
Set Membership: |
|

Genomic Map