Assay Details
Target Gene Details
Entrez Gene ID: | 69900 |
Gene Name: | major facilitator superfamily domain containing 11 |
Gene Aliases: |
2600014M03Rik |
Location: |
Chr.11:116852283-116875660 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd11 | NM_178620.3 | 13 | 2188 | NP_848735.1 |
| AK011216.1 | ||||
| AK031384.1 | 13 | 2383 | BAC27376.1 | |
| AK032043.1 | 13 | 2384 | BAC27666.1 | |
| BC088988.1 | 13 | 2188 | AAH88988.1 |

Genomic Map