Assay Details
Target Gene Details
Entrez Gene ID: | 215723 |
Gene Name: | major facilitator superfamily domain containing 6-like |
Gene Aliases: |
- |
Location: |
Chr.11:68556186-68558245 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd6l | NM_146004.1 | 1 | 1718 | NP_666116.1 |
| BC024997.1 | 1 | 1718 | AAH24997.1 |

Genomic Map